Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$159.90 - $633.45

Specifications

Antigen MBD1
Concentration Lot-Specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2910
SDP
Encompass
View Documents
Active Motif
39858
10 μL
Each for $159.90
Only null left
Add to Cart
 
50-199-2909
SDP
Encompass
View Documents
Active Motif
39857
100 μL
Each for $633.45
Only null left
Add to Cart
 
Description

Description

Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription from unmethylated promoters.
Specifications

Specifications

MBD1
Western Blot
Unconjugated
Rabbit
Human
Q9UIS9
4152
This MBD1 antibody was raised against a peptide within the C-terminal region of human MBD1.
Primary
-20°C, Avoid Freeze/Thaw Cycles
MBD1
Lot-Specific
Polyclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; Methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription
MBD1
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.