Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD1 Recombinant Rabbit Monoclonal Antibody (10B10)
GREENER_CHOICE

Catalog No. FEMA549992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549992 Supplier Invitrogen™ Supplier No. MA549992
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MBD1 Recombinant Monoclonal Antibody for ICC/IF, ELISA

Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription from unmethylated promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD1
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10B10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MBD1
Gene Accession No. Q9UIS9
Gene Alias CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription
Gene Symbols MBD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MBD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.