Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10055415
Description
MBD2 Polyclonal specifically detects MBD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
MBD2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to resognize isoform 1 (NP_003918.1) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_003918.1 | |
MBD2 | |
Peptide with sequence C-RNDPLNQNKGKPDLN corresponding to internal region according to NP_003918.1. | |
0.1 mg | |
Breast Cancer, Chromatin Research, Stem Cell Markers | |
8932 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction