Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD2 Antibody (SR1478), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321856100UL
Description
MBD2 Monoclonal antibody specifically detects MBD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
MBD2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1478 | |
Western Blot 1:5000-1:10000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41 | |
A synthesized peptide derived from human MBD2 (Uniprot #: Q9UBB5) | |
100 μg | |
Breast Cancer, Chromatin Research, Stem Cell Markers | |
8932 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction