Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD2 Rabbit anti-Mouse, Clone: 4D4D5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31631720UL
Description
MBD2 Monoclonal antibody specifically detects MBD2 in Mouse samples. It is validated for Western BlotSpecifications
| MBD2 | |
| Monoclonal | |
| Unconjugated | |
| demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MBD2 (Q9UBB5). | |
| 20 μg | |
| Breast Cancer, Chromatin Research, Stem Cell Markers | |
| 8932 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4D4D5 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction