Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD3 Antibody (PCRP-MBD3-1C4), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308535MFV450
Description
MBD3 Monoclonal antibody specifically detects MBD3 in Human samples. It is validated for Flow CytometrySpecifications
MBD3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-MBD3-1C4 | |
Flow Cytometry | |
methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3 | |
Recombinant full-length human MBD3 protein (Uniprot: O95983) | |
0.1 mL | |
Chromatin Research | |
53615 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction