Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534851 Supplier Invitrogen™ Supplier No. PA534851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431 nuclear. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (93%), Chicken (96%), Rhesus Monkey (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD3
Applications Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MBD3
Gene Accession No. O95983
Gene Alias AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3
Gene Symbols MBD3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MBD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53615
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.