Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD3L1 Monoclonal Antibody (1C7)
GREENER_CHOICE

Catalog No. PIMA524501
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524501 Supplier Invitrogen™ Supplier No. MA524501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAKSSQRKQR DCVNQCKSKP GLSTSIPLRM SSYTFKRPVT RITPHPGNEV RYHQWEESLE KPQQVCWQRR LQGLQAYSSA GELSSTLDLA NTLQKLVPSY TGGSLLEDL.

This gene encodes a protein that is related to methyl-CpG-binding proteins but lacks the methyl-CpG binding domain. The protein is localized to discrete areas in the nucleus, and expression appears to be restricted to round spermatids, suggesting that the protein plays a role in the postmeiotic stages of male germ cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD3L1
Applications ELISA
Classification Monoclonal
Clone 1C7
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MBD3L1
Gene Accession No. Q8WWY6
Gene Alias 1700070G05Rik; 1700095H13Rik; LOC100717039; Mbd3l; MBD3L1; MBD3-like protein 1; methyl-CpG binding domain protein 3 like 1; methyl-CpG binding domain protein 3-like 1; methyl-CpG-binding domain protein 3-like 1; Unknown (protein for MGC:137086)
Gene Symbols MBD3L1
Host Species Mouse
Immunogen MBD3L1 (NP_660209.1, 1 a.a. ~ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85509
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.