Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116154
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116154 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116154 Supplier Invitrogen™ Supplier No. PA5116154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MBD3L2 Polyclonal Antibody for Western Blot

This gene encodes a protein that is related to methyl-CpG-binding proteins but lacks the methyl-CpG binding domain. The protein has been found in germ cell tumors and some somatic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD3L2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MBD3L2
Gene Accession No. Q8NHZ7
Gene Alias CTB-25J19.1; MBD3L2; MBD3-like protein 2; methyl-CpG binding domain protein 3 like 2; methyl-CpG binding domain protein 3-like 2; methyl-CpG-binding domain protein 3-like 2
Gene Symbols MBD3L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 176-204 amino acids from the C-terminal region of human MBD3L2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125997
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.