Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ MBD4 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 8930 |
Concentration | 1 mg/ml |
Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-91189. For further blocking peptide related protocol, click here . |
Gene Alias | EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein |
Gene Symbol | MBD4 |
Quantity | 0.1 mg |
Research Category | Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction