Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565163 Supplier Invitrogen™ Supplier No. PA565163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGGGTNATPVV PSRAATPRSV RNKSHEGITN SVMPECKNPF KLMIGSSNAM GRLYVQELPG SQQQELHPVY PRQRLGSSEH GQKSPFRGSH Highest antigen sequence identity to the following orthologs - mouse 97%, rat 89%.

MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MBD5
Gene Accession No. Q9P267
Gene Alias 9430004D19Rik; AA536666; AI426407; C030040A15Rik; FLJ11113; FLJ30517; Gm1630; KIAA1461; MBD5; methyl-CpG binding domain protein 5; methyl-CpG-binding domain protein 5; Methyl-CpG-binding protein MBD5; MRD1; OTTMUSG00000012483
Gene Symbols MBD5
Host Species Rabbit
Immunogen Recombinant Human MBD5. Recombinant protein control fragment (Product #RP-105914).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.