Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MBD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578533
Description
Positive Control: HepG2, HepG2 nuclear extract Predicted Reactivity: Mouse (96%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
Specifications
| MBD5 | |
| Polyclonal | |
| Unconjugated | |
| MBD5 | |
| 9430004D19Rik; AA536666; AI426407; C030040A15Rik; FLJ11113; FLJ30517; Gm1630; KIAA1461; MBD5; methyl-CpG binding domain protein 5; methyl-CpG-binding domain protein 5; Methyl-CpG-binding protein MBD5; MRD1; OTTMUSG00000012483 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.19 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9P267 | |
| MBD5 | |
| Recombinant protein encompassing a sequence within the center region of human MBD5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction