Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MBD5 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0601
![]() ![]() |
Proteintech Group Inc
159611AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).Specifications
MBD5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
B1AYB6, Q9P267 | |
109241, 311026, 55777 | |
MBD5 Fusion Protein Ag8728 | |
Primary | |
-20°C | |
MBD5 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1461, MBD5, MRD1 | |
MBD5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title