Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD5 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABE132225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1322-25UG 25 μg
ABE1322-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE1322-25UG Supplier MilliporeSigma Supplier No. ABE132225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MBD5, Cat. No. ABE1322, is a highly specific rabbit polyclonal antibody that targets Methyl-CpG-binding domain protein 5 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD5
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Methyl-CpG-binding domain protein 5;Methyl-CpG-binding protein MBD
Gene Symbols MBD5;KIAA1461
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human Methyl-CpG-binding domain protein 5 (MBD5).
Molecular Weight of Antigen ∽160 kDa observed; 159.89 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060798.2
Test Specificity This rabbit polyclonal antibody detects isoform 1 of human Methyl-CpG-binding domain protein 5. It targets an epitope within 16 amino acids fromt the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.