Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116920 Supplier Invitrogen™ Supplier No. PA5116920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: PC-3, Mouse brain, Mouse thymus, Rat brain.

Binds to heterochromatin. Does not interact with either methylated or unmethylated DNA (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene MBD6
Gene Accession No. Q3TY92, Q96DN6
Gene Alias D10Wsu93e; KIAA1887; MBD6; methyl-CpG binding domain protein 6; methyl-CpG-binding domain protein 6; Methyl-CpG-binding protein MBD6
Gene Symbols MBD6
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110962, 114785, 362892
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.