Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBIP Mouse anti-Human, Clone: 2E6H11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MBIP |
---|---|
Clone | 2E6H11 |
Concentration | 1.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6538
![]() ![]() |
Proteintech Group Inc
661021IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.Specifications
MBIP | |
1.21 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MBIP, MUK binding inhibitory protein | |
MBIP | |
IgG2b | |
Protein A | |
Antibody |
2E6H11 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NS73 | |
51562 | |
MBIP Fusion Protein Ag18090 | |
Primary | |
-20°C | |
MBIP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title