Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103364 Supplier Invitrogen™ Supplier No. PA5103364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MBIP1.

MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MBIP
Gene Accession No. Q9NS73
Gene Alias 4933408E06Rik; BM-015; MAP3K12 binding inhibitory protein 1; MAP3K12-binding inhibitory protein 1; MAPK upstream kinase-binding inhibitory protein; MBIP; MUK-binding inhibitory protein; Unknown (protein for MGC:143421)
Gene Symbols MBIP
Host Species Rabbit
Immunogen A synthesized peptide derived from human MBIP(Accession Q9NS73), corresponding to amino acid residues R215-G265.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51562
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.