Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBL Antibody (3E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311223
Description
MBL Monoclonal specifically detects MBL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Frozen, Functional, Immunoassay.Specifications
MBL | |
Monoclonal | |
0.1 mg/ml | |
Flow Cytometry, Immunohistochemistry-Frozen, Functional, Immunoassay | |
COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833 | |
Serum isolated human RaRF | |
50 μg | |
Immunology | |
4153 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
30000000 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction