Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBL Antibody (OTI1A12), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272594AF594
Description
MBL Monoclonal specifically detects MBL in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MBL | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MBL2 | |
Full length recombinant protein of human MBL (NP_000233) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A12 | |
Western Blot, Immunohistochemistry | |
COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833 | |
Mouse | |
0.1 mL | |
4153 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction