Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBL1 Monoclonal Antibody (8G6)
GREENER_CHOICE

Catalog No. PIMA533352
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533352 Supplier Invitrogen™ Supplier No. MA533352
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Application Notes: Western Blot - A non-reduced sample treatment was used. The band sizes are 191, 263 and 316 kDa. IHC-F - Tissue sections were fixed in 4% PBS-buffered formaldehyde and pretreated with 2% hydrogen peroxide in methanol for 20 minutes at 4°C to quench endogenous peroxidases. Rat IgG2a was used as a negative control.

Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBL1
Applications ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8G6
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene MBD3
Gene Accession No. Q9Z2D8
Gene Alias AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3
Gene Symbols MBD3
Host Species Rat
Immunogen Purified mouse MBL-A.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17192
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.