Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBL2 Monoclonal Antibody (3E7), FITC, Invitrogen™

Catalog No. FERMA549066
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549066 Supplier Thermo Scientific Supplier No. MA549066

Mouse Monoclonal Antibody

MBL2 Monoclonal antibody specifically detects MBL2 in Human samples. It is validated for Flow Cytometry, Functional Assay Assay, Immunohistochemistry (Frozen), Inhibition Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBL2
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Inhibition Assays
Classification Monoclonal
Clone 3E7
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.02% sodium azide
Gene MBL2
Gene Accession No. P11226
Gene Alias 27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin
Host Species Mouse
Immunogen Serum isolated human RaRF
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4153
Target Species Human
Content And Storage 4° C, store in dark
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.