Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110650 Supplier Invitrogen™ Supplier No. PA5110650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGDSSLAASE RKALQTEMAR IKKWLTFSLG KQVGNKFFLT NGEIMTFEKV KALCVKFQAS VATPRNAAEN GAIQNLIKEE AFLGITDEKT EGQFVDLTGN RLTYTNWNEG EPNNAGSDED CVLLLKNGQW NDVPCSTSH Highest antigen sequence indentity to the following orthologs: Mouse - 56%, Rat - 54%.

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MBL2
Gene Accession No. P11226
Gene Alias 27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin
Gene Symbols MBL2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MBL2. Recombinant protein control fragment (Product #RP-95395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.