Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBLAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564468 Supplier Invitrogen™ Supplier No. PA564468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDFCLPGGRY LPHGLGEGQP LRLGPGLEVW ATPGHGGQRD VSVVVAGTAL GTVVVAGDVF ERDGDE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

MBLAC1 gene ontology annotations related to this gene include hydrolase activity; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBLAC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MBLAC1
Gene Accession No. A4D2B0
Gene Alias MBLAC1; metallo-beta-lactamase domain containing 1; metallo-beta-lactamase domain-containing protein 1; RGD1306932; Unknown (protein for MGC:137924)
Gene Symbols MBLAC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MBLAC1. Recombinant protein control fragment (Product #RP-104029).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255374
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.