Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBLAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118052 Supplier Invitrogen™ Supplier No. PA5118052
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBLAC2 Polyclonal Antibody for Western Blot, IP

MBLAC2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBLAC2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene MBLAC2
Gene Accession No. Q68D91
Gene Alias MBLAC2; metallo-beta-lactamase domain containing 2; metallo-beta-lactamase domain-containing protein 2
Gene Symbols MBLAC2
Host Species Rabbit
Immunogen E. coli-derived Human MBLAC2 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153364
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.