Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBNL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287783
Description
MBNL2 Polyclonal specifically detects MBNL2 in Rat samples. It is validated for Western Blot.Specifications
MBNL2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
10150 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
DKFZp781H1296, MBLL39Muscleblind-like protein 1, MBLLMGC120628, MGC120625, MGC120626, MLP1, muscleblind-like 2 (Drosophila), muscleblind-like protein 2, Muscleblind-like protein-like, Muscleblind-like protein-like 39, PRO2032 | |
The immunogen is a synthetic peptide directed towards the middle region of RAT MBNL2 | |
100 μL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction