Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBNL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543698 Supplier Invitrogen™ Supplier No. PA543698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ENGRVIACFD SLKGRCSREN CKYLHPPTHL KTQLEINGRN NLIQQKTAAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene encodes a C3H-type zinc finger protein, which is similar to the Drosophila melanogaster muscle blind B protein. Drosophila muscle blind is a gene required for photoreceptor differentiation. Several alternatively spliced transcript variants have been described but the full-length natures of only some have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBNL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MBNL2
Gene Accession No. Q5VZF2
Gene Alias 1110002M11Rik; AI047808; AI837313; AI849185; AL118326; DKFZp781H1296; fj89c04; Kiaa4072; MBLL; MBLL39; mbnl2; MGC120625; MGC120626; MGC120628; mKIAA4072; MLP1; muscleblind like splicing regulator 2; muscleblind-like 2; muscleblind-like protein 1; Muscleblind-like protein 2; Muscleblind-like protein-like; Muscleblind-like protein-like 39; muscleblind-like splicing regulator 2; PRO2032; R75232; RP11-128N14.1; wu:fj89c04
Gene Symbols MBNL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MBNL2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.