Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBNL3 Antibody (PCRP-MBNL3-1D11), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308663AF350
Description
MBNL3 Monoclonal antibody specifically detects MBNL3 in Human samples. It is validated for Flow Cytometry, Protein ArraySpecifications
MBNL3 | |
Monoclonal | |
Alexa Fluor 350 | |
CHCRMBLX39FLJ11316, Cys3His CCG1-required protein, FLJ97142, MBXLMBLX, muscleblind-like 3 (Drosophila), muscleblind-like protein 3, Muscleblind-like X-linked protein, Protein HCHCR | |
Recombinant full-length human MBNL3 protein (Uniprot: Q9NUK0) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Protein Array | |
PCRP-MBNL3-1D11 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
55796 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction