Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MBNL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550445
Description
The antibody detects endogenous levels of total MBNL3 protein.
This gene encodes a member of the muscleblind-like family of proteins. The encoded protein may function in regulation of alternative splicing and may play a role in the pathophysiology of myotonic dystrophy. Alternatively spliced transcript variants have been described.
Specifications
MBNL3 | |
Polyclonal | |
Unconjugated | |
Mbnl3 | |
A530038J18Rik; AI661274; CHCR; Cys3His CCG1-required protein; E430034C16Rik; MBLX; MBLX39; Mbnl3; MBXL; muscleblind like splicing regulator 3; muscleblind-like 3; muscleblind-like 3 (Drosophila); muscleblind-like protein 3; muscleblind-like splicing regulator 3; muscleblind-like X-linked protein; Protein HCHCR; Protein MCHCR; RGD1565834; RP5-842K24.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55796 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NUK0 | |
Mbnl3 | |
Fusion protein corresponding to a region derived from internal residues of human muscleblind-like splicing regulator 3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction