Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBNL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986749
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6749 Supplier Bioss Supplier No. BS5721R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the muscleblind-like family of proteins. The encoded protein may function in regulation of alternative splicing and may play a role in the pathophysiology of myotonic dystrophy. Alternatively spliced transcript variants have been described.

Specifications

Antigen MBNL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mbnl3
Gene Accession No. Q9NUK0
Gene Alias A530038J18Rik; AI661274; Chcr; Cys3His CCG1-required protein; E430034C16Rik; MBLX; MBLX39; Mbnl3; Mbxl; muscleblind like splicing regulator 3; muscleblind-like 3; muscleblind-like 3 (Drosophila); Muscleblind-like protein 3; muscleblind-like splicing regulator 3; Muscleblind-like X-linked protein; Protein HCHCR; Protein MCHCR; RP5-842K24.3
Gene Symbols Mbnl3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MBNL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55796
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.