Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MBOAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553238
Description
Immunogen sequence: TQRRKNTHEN IQLSQSKKFD EGENSLGQNS FSTTNNVCNQ NQEIASRHSS LKQ Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
Acyltransferase which mediates the conversion of lysophosphatidylethanolamine (1-acyl-sn-glycero-3- phosphoethanolamine or LPE) into phosphatidylethanolamine (1,2- diacyl-sn-glycero-3-phosphoethanolamine or PE) (LPEAT activity). Catalyzes also the acylation of lysophosphatidic acid (LPA) into phosphatidic acid (PA) (LPAAT activity). Has also a very weak lysophosphatidylcholine acyltransferase (LPCAT activity). Prefers oleoyl-CoA as the acyl donor. Lysophospholipid acyltransferases (LPLATs) catalyze the reacylation step of the phospholipid remodeling pathway also known as the Lands cycle.
Specifications
MBOAT2 | |
Polyclonal | |
Unconjugated | |
MBOAT2 | |
1-acylglycerophosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 231n7; 2810049G06Rik; AU022889; AW547221; LPAAT; LPC acyltransferase 4; LPCAT; Lpcat4; LPEAT; LPLAT 2; lyso-PA acyltransferase; lyso-PC acyltransferase 4; lyso-PE acyltransferase; lysophosphatidic acid acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase; Lysophospholipid acyltransferase 2; MBOAT2; membrane bound O-acyltransferase domain containing 2; membrane-bound O-acyltransferase domain-containing protein 2; mLPCAT4; Moact2; OACT2; O-acyltransferase (membrane bound) domain containing 2; O-acyltransferase domain-containing protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
129642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZWT7 | |
MBOAT2 | |
Recombinant protein corresponding to Human MBOAT2. Recombinant protein control fragment (Product #RP-91250). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction