Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569582 Supplier Invitrogen™ Supplier No. PA569582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.

Acyltransferase which mediates the conversion of lysophosphatidylethanolamine (1-acyl-sn-glycero-3- phosphoethanolamine or LPE) into phosphatidylethanolamine (1,2- diacyl-sn-glycero-3-phosphoethanolamine or PE) (LPEAT activity). Catalyzes also the acylation of lysophosphatidic acid (LPA) into phosphatidic acid (PA) (LPAAT activity). Has also a very weak lysophosphatidylcholine acyltransferase (LPCAT activity). Prefers oleoyl-CoA as the acyl donor. Lysophospholipid acyltransferases (LPLATs) catalyze the reacylation step of the phospholipid remodeling pathway also known as the Lands cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MBOAT2
Gene Accession No. Q3T1J2
Gene Alias 1-acylglycerophosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 231n7; 2810049G06Rik; AU022889; AW547221; LPAAT; LPC acyltransferase 4; LPCAT; Lpcat4; LPEAT; LPLAT 2; lyso-PA acyltransferase; lyso-PC acyltransferase 4; lyso-PE acyltransferase; lysophosphatidic acid acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase; Lysophospholipid acyltransferase 2; MBOAT2; membrane bound O-acyltransferase domain containing 2; membrane-bound O-acyltransferase domain-containing protein 2; mLPCAT4; Moact2; OACT2; O-acyltransferase (membrane bound) domain containing 2; O-acyltransferase domain-containing protein 2
Gene Symbols MBOAT2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat MBOAT2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313997
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.