Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545952 Supplier Invitrogen™ Supplier No. PA545952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FKLTYYSHWI LDDSLLHAAG FGPELGQSPG EEGYVPDADI WTLERTHRIS Sequence homology: Cow: 85%; Dog: 85%; Horse: 79%; Human: 100%; Mouse: 79%; Sheep: 77%.

Mediates the octanoylation of ghrelin at 'Ser-3'. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MBOAT4
Gene Accession No. Q96T53
Gene Alias EMBL:ACB05875.1}; FKSG89; Ghrelin O-acyltransferase; ghrelin O-acyltransferase {ECO:0000303; Gm171; GOAT; Mboat4; mboat4 {ECO:0000312; membrane bound O-acyltransferase domain containing 4; membrane-bound O-acyltransferase domain-containing protein 4; membrane-bound O-acyltransferase domain-containing protein 4 {ECO:0000312; OACT4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4; PubMed:18443287}; RGD:1566335}; RGD1566335
Gene Symbols MBOAT4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MBOAT4 (aa 251-300).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 619373
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.