Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532916
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532916 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532916 Supplier Invitrogen™ Supplier No. PA532916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan (100%) Gibbon, Monkey, Galago, Panda, Rabbit (93%) Mouse, Bat, Horse (86%).

Mediates the octanoylation of ghrelin at 'Ser-3'. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MBOAT4
Gene Accession No. Q96T53
Gene Alias EMBL:ACB05875.1}; FKSG89; Ghrelin O-acyltransferase; ghrelin O-acyltransferase {ECO:0000303; Gm171; GOAT; Mboat4; mboat4 {ECO:0000312; membrane bound O-acyltransferase domain containing 4; membrane-bound O-acyltransferase domain-containing protein 4; membrane-bound O-acyltransferase domain-containing protein 4 {ECO:0000312; OACT4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4; PubMed:18443287}; RGD:1566335}; RGD1566335
Gene Symbols MBOAT4
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from internal region of human MBOAT4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 619373
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.