Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBOAT7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169610
Description
MBOAT7 Polyclonal specifically detects MBOAT7 in Human samples. It is validated for Western Blot.Specifications
| MBOAT7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 1-acylglycerophosphatidylinositol O-acyltransferase, BB1LPLAT 7, Bladder and breast carcinoma-overexpressed gene 1 protein, EC 2.3.1.-, EC 2.3.1.n4, h-mboa-7, hMBOA-7, LENG4FLJ41296, leukocyte receptor cluster (LRC) member 4, Leukocyte receptor cluster member 4, LPIATO-acyltransferase domain-containing protein 7, LRC4, Lysophosphatidylinositol acyltransferase, lysophospholipid acyltransferase 7, Lyso-PI acyltransferase, malignant cell expression-enhanced gene/tumor progression-enhanced, MBOA7, membrane bound O-acyltransferase domain containing 7, Membrane-bound O-acyltransferase domain-containing protein 7, OACT7 | |
| Rabbit | |
| 53 kDa | |
| 100 μL | |
| Primary | |
| Rat: 86%; Mouse: 86%. | |
| Human, Mouse, Rat, Pig, Canine, Equine | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96N66 | |
| MBOAT7 | |
| Synthetic peptides corresponding to LENG4 The peptide sequence was selected from the C terminal of LENG4. Peptide sequence LSAYWHGLHPGYYLSFLTIPLCLAAEGRLESALRGRLSPGGQKAWDWVHW. | |
| Protein A purified | |
| RUO | |
| 79143 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction