Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBOAT7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | MBOAT7 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MBOAT7 Polyclonal antibody specifically detects MBOAT7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MBOAT7 | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
1-acylglycerophosphatidylinositol O-acyltransferase, BB1LPLAT 7, Bladder and breast carcinoma-overexpressed gene 1 protein, EC 2.3.1.-, EC 2.3.1.n4, h-mboa-7, hMBOA-7, LENG4FLJ41296, leukocyte receptor cluster (LRC) member 4, Leukocyte receptor cluster member 4, LPIATO-acyltransferase domain-containing protein 7, LRC4, Lysophosphatidylinositol acyltransferase, lysophospholipid acyltransferase 7, Lyso-PI acyltransferase, malignant cell expression-enhanced gene/tumor progression-enhanced, MBOA7, membrane bound O-acyltransferase domain containing 7, Membrane-bound O-acyltransferase domain-containing protein 7, OACT7 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LGGGSPSRRKAASQPTSLAPEKLREE | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2), 40% Glycerol | |
79143 | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title