Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563224 Supplier Invitrogen™ Supplier No. PA563224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGGGSPSRRK AASQPTSLAP EKLREE Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

MBOAT7 belongs to the membrane-bound acyltransferase family. It is involved in arachidonate recycling, thus regulating free arachidonic acid levels and leukotriene synthesis in neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MBOAT7
Gene Accession No. Q96N66
Gene Alias 1-acylglycerophosphatidylinositol O-acyltransferase; 5730589L02Rik; BB1; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; hMBOA-7; Leng4; leukocyte receptor cluster (LRC) member 4; leukocyte receptor cluster member 4; LPIAT; Lpiat1; LPLAT 7; LRC4; lysophosphatidylinositol acyltransferase; Lysophosphatidylinositol acyltransferase 1; Lysophospholipid acyltransferase 7; lyso-PI acyltransferase; malignant cell expression-enhanced gene/tumor progression-enhanced; mBB1; MBOA7; Mboat7; membrane bound O-acyltransferase domain containing 7; membrane-bound O-acyltransferase domain-containing protein 7; m-mboa-7; OACT7; O-acyltransferase domain-containing protein 7; RGD1306945
Gene Symbols MBOAT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MBOAT7. Recombinant protein control fragment (Product #RP-101661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.