Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543430 Supplier Invitrogen™ Supplier No. PA543430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WWLAQYIYKS APARSYVLRS AWTMLLSAYW HGLHPGYYLS FLTIPLCLAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.

MBOAT7 belongs to the membrane-bound acyltransferase family. It is involved in arachidonate recycling, thus regulating free arachidonic acid levels and leukotriene synthesis in neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MBOAT7
Gene Accession No. Q96N66
Gene Alias 1-acylglycerophosphatidylinositol O-acyltransferase; 5730589L02Rik; BB1; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; hMBOA-7; Leng4; leukocyte receptor cluster (LRC) member 4; leukocyte receptor cluster member 4; LPIAT; Lpiat1; LPLAT 7; LRC4; lysophosphatidylinositol acyltransferase; Lysophosphatidylinositol acyltransferase 1; Lysophospholipid acyltransferase 7; lyso-PI acyltransferase; malignant cell expression-enhanced gene/tumor progression-enhanced; mBB1; MBOA7; Mboat7; membrane bound O-acyltransferase domain containing 7; membrane-bound O-acyltransferase domain-containing protein 7; m-mboa-7; OACT7; O-acyltransferase domain-containing protein 7; RGD1306945
Gene Symbols MBOAT7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LENG4 (aa 326-375).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.