Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Antibody (2H9), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222121MFV450
Description
MBP Monoclonal antibody specifically detects MBP in Human, Mouse samples. It is validated for Flow CytometrySpecifications
MBP | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
2H9 | |
Flow Cytometry | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
Purified recombinant fragment of human Myelin Basic Protein expressed in E. coli. [UniProt# P02686] | |
0.1 mL | |
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
4155 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction