Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Antibody (HL1033), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP313687
This item is not returnable.
View return policy
Description
MBP Monoclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| MBP | |
| Monoclonal | |
| Unconjugated | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse MBP. The exact sequence is proprietary. | |
| 100 μg | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HL1033 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction