Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Antibody, Novus Biologicals™
SDP

Catalog No. NBP229656 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229656 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229656 Supplier Novus Biologicals Supplier No. NBP229656
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MBP Polyclonal specifically detects MBP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MBP
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500
Formulation PBS with 10mM Sodium Azide
Gene Accession No. P02688
Gene Alias MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Gene Symbols MBP
Host Species Chicken
Immunogen Three peptide sequences conserved in higher vertebrate MBP proteins. Accession # P02688
Molecular Weight of Antigen 18 kDa
Purification Method IgY purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4155
Test Specificity Specific for the ∼18k & 22k myelin basic protein in Western blots of rat brain homogenate.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.