Learn More
Thermo Scientific™ MBP Column Preparation Buffer for Pierce™ IgM Purification Kit PROMO
Catalog No. PI21018PM
Click to view available options
:
Thermo Scientific™ MBP Column Preparation Buffer for Pierce™ IgM Purification Kit
Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5 mL column of the MBP Agarose.
Highlights:
  • Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum
  • Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5 mL ascites samples per column-use
  • Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies
  • Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity
Includes:
Kit contains a 5 mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender
Specifications
Product Type IgM Purification Kit
Product Line Pierce™
Reagent Type Conditioning Reagent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C.
Quantity 50 mL
Form Liquid
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.