Learn More
Invitrogen™ MBP Monoclonal Antibody (CL2827)

Description
Immunogen sequence: DENPVVHFFK NIVTPRTPPP SQGKGRGLSL SRFSWGAEGQ RPGFGYGGRA SDYKSAHKGF KGVDAQGTLS KIFKLGGRDS RSGSPM Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.
Specifications
Specifications
| Antigen | MBP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL2827 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MBP |
| Gene Accession No. | P02686, P02688, P04370 |
| Gene Alias | 20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.