Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-577 Supplier Invitrogen™ Supplier No. PA118324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called Golli-MBP) that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBP
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene MBP
Gene Accession No. P02686, P02688, P25188
Gene Alias 20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product
Gene Symbols MBP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues Y(68) G S L P Q K S Q R S Q D E N P V V(86) of guinea pig MBP, conjugated to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100731253, 24547, 4155
Target Species Human, Rat, Guinea Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.