Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597975
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597975 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597975 Supplier Invitrogen™ Supplier No. PA597975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOBP Polyclonal Antibody for Western Blot, ICC/IF, ELISA

MOBP may play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOBP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Mobp
Gene Accession No. Q13875
Gene Alias AI835053; AI839848; Mobp; MOBP155; MOBP170; MOBP69; MOBP73; Mobp81; Mobp81p; myelin associated oligodendrocyte basic protein; myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; R74645; RNMOBP81P; Unknown (protein for MGC:128384)
Gene Symbols Mobp
Host Species Rabbit
Immunogen Recombinant Human Myelin-associated oligodendrocyte basic protein (1-81AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4336
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.