Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102458 Supplier Thermo Scientific Supplier No. PA5102458
Add to Cart
Add to Cart

MBP Polyclonal Antibody

MBP Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MBP
Gene Accession No. P02686, P02688, P04370
Gene Alias 20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product
Gene Symbols MBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human MBP(Accession P02686), corresponding to amino acid residues L206-R256.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17196, 24547, 4155
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.