Learn More
Description
- Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
- Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
- Maximum pressure: 3 bar
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
- 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Buffer | Aqueous Buffer |
| Product Type | Dextrin Sepharose HP prepacked column |
| Bed Volume | 1mL |
| Diameter (English) Inner | 0.27 in. |
| Diameter (Metric) Inner | 7mm |
| Quantity | 1 x 1mL |
| Height (English) | 0.98 in. |
| Height (Metric) | 25mm |
| Flow Rate | <4 mL/min. |
| Material | Polypropylene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.