Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543599 Supplier Invitrogen™ Supplier No. PA543599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PFKPCMRVEV VDKRHLCRTR VAVVESVIGG RLRLVYEESE DRTDDFWCHM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 86%.

Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. Specifically binds to monomethylated and dimethylated 'Lys-20' on histone H4. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBTD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mbtd1
Gene Accession No. Q05BQ5
Gene Alias AA408199; AI194990; hemp; im:7159307; mbt domain containing 1; MBT domain-containing protein 1; MBT domain-containing protein 1; LOW QUALITY PROTEIN: MBT domain-containing protein 1; MBTD1; SA49P01
Gene Symbols Mbtd1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MBTD1 (aa 284-333).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54799
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.