Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MBTFA Acylation Reagent

Catalog No. PI49704
Change view
Click to view available options
Quantity:
10 x 1 mL ampules
100 mL
25 mL
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI49704 100 mL
PI49700 10 x 1 mL ampules
TS49701 5 g
PI49703 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI49704 Supplier Thermo Scientific™ Supplier No. TS49704

Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent.

Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent. The principal byproduct N-methyltrifluoroacetamine is stable, volatile, and does not present problems in subsequent GC. MBTFA produces very volatile derivatives of carbohydrates and can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation.

  • Reacts under nonacidic conditions
  • Principle byproduct from the derivatization reaction is N-methyltrifluoroacetamine, which is stable, volatile and does not present problems in subsequent GC
  • Produces very volatile derivatives of carbohydrates
  • Can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Standard Components N-Methyl-bis(trifluoroacetamide)
Quantity 100 mL
Storage -20°C
Hazards Corrosive, Flammable
Standard Concentration 98 to 100%
Item Description 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.