Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTPS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | MBTPS1 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MBTPS1 Polyclonal antibody specifically detects MBTPS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MBTPS1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Endocrinology, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
8720 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.4.21.112, Endopeptidase S1P, KIAA0091MGC138712, membrane-bound transcription factor peptidase, site 1, membrane-bound transcription factor protease, site 1, PCSK8, S1PMGC138711, site-1 protease, SKI1, SKI-1membrane-bound transcription factor site-1 protease, subtilisin/kexin isozyme-1, Subtilisin/kexin-isozyme 1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: YPPGYFPRDNLRMKNDPLDWNGDHIHTNFRDMYQHLRSMGYFVEVLGAPFTCFDASQYGTLLMVDSEEEYFPEEIAKLRRDVDNGLSLVIFSDWYNTSVMRKVKFYDENTRQWWMPDTGGANIPALNELLSV | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title