Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBTPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563247 Supplier Invitrogen™ Supplier No. PA563247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPPGYFPRDN LRMKNDPLDW NGDHIHTNFR DMYQHLRSMG YFVEVLGAPF TCFDASQYGT LLMVDSEEEY FPEEIAKLRR DVDNGLSLVI FSDWYNTSVM RKVKFYDENT RQWWMPDTGG ANIPALNELL SV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

This gene encodes a member of the subtilisin-like proprotein convertase family, which includes proteases that process protein and peptide precursors trafficking through regulated or constitutive branches of the secretory pathway. The encoded protein undergoes an initial autocatalytic processing event in the ER to generate a heterodimer which exits the ER and sorts to the cis/medial-Golgi where a second autocatalytic event takes place and the catalytic activity is acquired. It encodes a type 1 membrane bound protease which is ubiquitously expressed and regulates cholesterol or lipid homeostasis via cleavage of substrates at non-basic residues. Mutations in this gene may be associated with lysosomal dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBTPS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MBTPS1
Gene Accession No. Q14703
Gene Alias 0610038M03Rik; AV003995; endopeptidase S1P; KIAA0091; MBTPS1; membrane bound transcription factor peptidase, site 1; membrane-bound transcription factor peptidase, site 1; membrane-bound transcription factor protease, site 1; membrane-bound transcription factor site-1 protease; mKIAA0091; PCSK8; proprotein convertase subtilisin/kexin type 8; S1p; S1P endopeptidase; site-1 protease; Ski1; SKI-1; Sterol-regulated luminal protease; subtilisin kexin isozyme 1; subtilisin/kexin isozyme 1; subtilisin/kexin isozyme SKI-1; subtilisin/kexin isozyme-1; Subtilisin/kexin-isozyme 1
Gene Symbols MBTPS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MBTPS1. Recombinant protein control fragment (Product #RP-104051).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.